Learn More
Teledyne ISCO, Inc Ls Class Single Piston Pump With Belt Drive And Shaped Cam; 0.000 - 5.000 Ml/Min.; 6,000 Psi
Catalog No. 502119946 Shop All Teledyne ISCO Inc. Products
Encompass
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.000 - 5.000 mL/min.; 6,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9946 Supplier Teledyne ISCO, Inc Supplier No. LS005SRX
Add to Cart
Add to Cart
Request a Quote
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.000 - 5.000 mL/min.; 6,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing