Learn More
Kuhner Shaker Inc LS-Z SHAKER AMBIENT TEMP
Catalog No. 502569940
Encompass
LS-Z ShakerAmbient temperature: -20CC to 80CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel housingShaker DIMS: H189 x W526 x D550mmPower Supply DIMS: H109 x W133 x D428mmController DIMS: H97 x W220 x D x 150mmTotal Weight: 40kgFIVE YEAR WARRANTY HARDWARE
Catalog No. 50-256-9940 Supplier Kuhner Shaker Inc Supplier No. SMZ1200
Add to Cart
Add to Cart
Request a Quote
LS-Z ShakerAmbient temperature: -20CC to 80CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel housingShaker DIMS: H189 x W526 x D550mmPower Supply DIMS: H109 x W133 x D428mmController DIMS: H97 x W220 x D x 150mmTotal Weight: 40kgFIVE YEAR WARRANTY HARDWARE