Learn More
ATCC LS123; Colon Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL255
Encompass
LS123 is an epithelial cell line that was isolated in 1974 from the colon of a White, 65-year-old, female patient with colorectal adenocarcinoma. This cell line was deposited by LP Rutzky and, can be used in cancer research.
Catalog No. 50-238-2462