Learn More
ATCC LS411N; Colorectal Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2159
Encompass
LS411N is a cell line exhibiting epithelial morphology that was isolated from the large intestine (cecum) of a patient with Dukes' type B carcinoma. It has applications in cancer research.
Catalog No. 50-238-2609
 Restricted Item