Learn More
Labrepco Inc LS4800 LABSYSTEM (W/ RACKS W/
SDP
Catalog No. 509020688
Encompass_Preferred
Storage unit, Cryogenic sample; Labrepco; With 6 racks; Does not include boxes (holds 5 x 5 in. 100 vial boxes); Capacity: 4800 x 2.0mL vials; Overall height: 35.1 in.; Overall dia.: 26.9 in.
Catalog No. 50-902-0688 Supplier Labrepco Inc Supplier No. LS4800
Add to Cart
Add to Cart
Request a Quote
Storage unit, Cryogenic sample; Labrepco; With 6 racks; Does not include boxes (holds 5 x 5 in. 100 vial boxes); Capacity: 4800 x 2.0mL vials; Overall height: 35.1 in.; Overall dia.: 26.9 in.