Learn More
Labrepco Inc LS750 LABSYSTEM (W/ RACKS W/O
SDP
Catalog No. 509020678
Encompass_Preferred
Storage unit, Cryogenic sample; Labrepco; With 6 racks; Does not include boxes (holds 3 x 3 in. 25 vial boxes); Capacity: 750 x 2.0mL vials; Overall height: 26.8 in.; Overall dia.: 18.8 in.
Catalog No. 50-902-0678 Supplier Labrepco Inc Supplier No. LS750BR
Storage unit, Cryogenic sample; Labrepco; With 6 racks; Does not include boxes (holds 3 x 3 in. 25 vial boxes); Capacity: 750 x 2.0mL vials; Overall height: 26.8 in.; Overall dia.: 18.8 in.