Learn More
Aviva Systems Biology Corp LSAMP Antibody - N-terminal region (ARP45277_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45277_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSAMP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LSAMP. Alternate names: LAMP, IGLON3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58997