Learn More
Medchemexpress LLC LSD (acetate) | 945245-76-1 | 98.0% | 1009.20 | 10 MG
SDP
Catalog No. 5000399454
Encompass_Preferred
LSD acetate is a peptide utilized in targeted therapy and diagnosis research for tumor lymphatics. It specifically recognizes the lymphatics of C8161 melanoma, without binding to normal tissue lymphatics or tumor blood vessels. When conjugated with a proapoptotic peptide, it can effectively reduce the number of tumor lymphatics.
  • Recognizes C8161 melanoma lymphatics
  • Does not bind to normal tissue lymphatics or tumor blood vessels
  • Reduces tumor lymphatics when conjugated with proapoptotic peptides
  • Suitable for targeted therapy research
  • Suitable for diagnosis of tumor lymphatics
Catalog No. 50-003-99454 Supplier Medchemexpress LLC Supplier No. HYP4091A10MG
LSD acetate is a peptide utilized in targeted therapy and diagnosis research for tumor lymphatics. It specifically recognizes the lymphatics of C8161 melanoma, without binding to normal tissue lymphatics or tumor blood vessels. When conjugated with a proapoptotic peptide, it can effectively reduce the number of tumor lymphatics.
  • Recognizes C8161 melanoma lymphatics
  • Does not bind to normal tissue lymphatics or tumor blood vessels
  • Reduces tumor lymphatics when conjugated with proapoptotic peptides
  • Suitable for targeted therapy research
  • Suitable for diagnosis of tumor lymphatics