Learn More
Medchemexpress LLC LSD acetate | 98.0% | 1009.20 | 25 MG
SDP
Catalog No. 5000396207
Encompass_Preferred
LSD acetate is a peptide that specifically recognizes the lymphatics of C8161 melanoma, but not the lymphatics of normal tissues or tumor blood vessels. When conjugated with a proapoptotic peptide, it can reduce the number of tumor lymphatics. It can be used in the research of targeted therapy and diagnosis of tumor lymphatics.
  • Specifically recognizes lymphatics of C8161 melanoma.
  • Does not bind to lymphatics of normal tissues or tumor blood vessels.
  • Can reduce the number of tumor lymphatics when conjugated with a proapoptotic peptide.
  • Useful in research for targeted therapy and diagnosis of tumor lymphatics.
Catalog No. 50-003-96207 Supplier Medchemexpress LLC Supplier No. 25MGHYP4091A
LSD acetate is a peptide that specifically recognizes the lymphatics of C8161 melanoma, but not the lymphatics of normal tissues or tumor blood vessels. When conjugated with a proapoptotic peptide, it can reduce the number of tumor lymphatics. It can be used in the research of targeted therapy and diagnosis of tumor lymphatics.
  • Specifically recognizes lymphatics of C8161 melanoma.
  • Does not bind to lymphatics of normal tissues or tumor blood vessels.
  • Can reduce the number of tumor lymphatics when conjugated with a proapoptotic peptide.
  • Useful in research for targeted therapy and diagnosis of tumor lymphatics.