Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LSD acetate | 98.0% | 1009.20 (free base) | 5 MG
SDP

Catalog No. 5000398533
Encompass_Preferred

LSD acetate is a peptide designed for targeted research in oncology. It specifically recognizes the lymphatics of C8161 melanoma, distinguishing them from normal tissues and tumor blood vessels. This peptide can reduce the number of tumor lymphatics when conjugated with a proapoptotic peptide, making it valuable for the development of targeted therapies and diagnostic tools for tumor lymphatics.

  • Specifically recognizes lymphatics of C8161 melanoma
  • Does not bind to lymphatics of normal tissues or tumor blood vessels
  • Reduces tumor lymphatics when conjugated with a proapoptotic peptide
  • Useful for targeted therapy and diagnosis of tumor lymphatics research
  • Molecular weight: 1009.20 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Cys-Leu-Ser-Asp-Gly-Lys-Arg-Lys-Cys
  • Soluble in DMSO: 100 mg/mL (needs ultrasonic)

Catalog No. 50-003-98533 Supplier Medchemexpress LLC Supplier No. HYP4091A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LSD acetate is a peptide designed for targeted research in oncology. It specifically recognizes the lymphatics of C8161 melanoma, distinguishing them from normal tissues and tumor blood vessels. This peptide can reduce the number of tumor lymphatics when conjugated with a proapoptotic peptide, making it valuable for the development of targeted therapies and diagnostic tools for tumor lymphatics.

  • Specifically recognizes lymphatics of C8161 melanoma
  • Does not bind to lymphatics of normal tissues or tumor blood vessels
  • Reduces tumor lymphatics when conjugated with a proapoptotic peptide
  • Useful for targeted therapy and diagnosis of tumor lymphatics research
  • Molecular weight: 1009.20 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Cys-Leu-Ser-Asp-Gly-Lys-Arg-Lys-Cys
  • Soluble in DMSO: 100 mg/mL (needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.