Learn More
Epigentek Inc Epigenase LSD1 Demethylase Activity/Inhibition Assay Kit (Colorimetric) (48 assays)
SDP
Catalog No. 501096311
Encompass_Preferred
The Epigenase LSD1 Demethylase Activity/Inhibition Assay Kit (Colorimetric) is a complete set of optimized reagents, designed for an easy and fast colorimetric measurement of LSD1 activity or inhibition. The antibody-based, immunospecific method directly detects LSD1-converted demethylated products, rather than by-products, in a 96 stripwell microplate format.
Catalog No. 50-109-6311 Supplier Epigentek Inc Supplier No. P307848
The Epigenase LSD1 Demethylase Activity/Inhibition Assay Kit (Colorimetric) is a complete set of optimized reagents, designed for an easy and fast colorimetric measurement of LSD1 activity or inhibition. The antibody-based, immunospecific method directly detects LSD1-converted demethylated products, rather than by-products, in a 96 stripwell microplate format.