Learn More
Enzo Life Sciences LSD1 (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5440050
Encompass_Preferred
Alternative name: KDM1, Lysine specific histone demethylase 1, BHC110, AOF2, KIAA0601. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 8.1mM Na2HPO4, 1.5mM KH2PO4, pH 7.2, 138mM sodium chloride, 2.7mM KCl and 40% v/v glycerol. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2079