Learn More
BPS Bioscience Inc LSD1 (KDM1A) Homogeneous Assay Kit, 384 reactions
SDP
Catalog No. NC4036035
Encompass
The LSD1 (KDM1A) Homogenous Assay Kit is designed to measure activity of the LSD1 (lysine-specific histone demethylase 1A, also known as KDM1A) for screening and profiling purposes. The kit comes in a convenient format 384-well AlphaLISA® format, with enough purified recombinant LSD1 (amino acids 158-852(end)), primary antibody, substrate and buffers for 400 enzyme reactions.
Catalog No. NC4036035 Supplier BPS Bioscience Inc Supplier No. 50108
The LSD1 (KDM1A) Homogenous Assay Kit is designed to measure activity of the LSD1 (lysine-specific histone demethylase 1A, also known as KDM1A) for screening and profiling purposes. The kit comes in a convenient format 384-well AlphaLISA® format, with enough purified recombinant LSD1 (amino acids 158-852(end)), primary antibody, substrate and buffers for 400 enzyme reactions.