Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ LSE™ Shaking Incubator Accessory, Stacking Adapter

Catalog No. 07202173 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
LSE Shaking Incubator, 49L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-202-173 LSE Shaking Incubator, 49L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-202-173 Supplier Corning™ Supplier No. 480158
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with LSE Shaking Incubator

For use with Cat. Nos. 6750 and 6752.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stacking Adapter
For Use With (Equipment) LSE Shaking Incubator, 49L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.