Learn More
Cayman Chemical LSelectInCD62L EXrAClula 100ug
SDP
Catalog No. 502911143
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged mouse L-selectin expressed in HEK293 cells • Amino acids: 39-332 • MW: 61 kDa
Catalog No. 50-291-1143 Supplier Cayman Chemical Supplier No. 35727100 UG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged mouse L-selectin expressed in HEK293 cells • Amino acids: 39-332 • MW: 61 kDa