Learn More
Cayman Chemical LSelectInCD62L EXrAClula 200ug
SDP
Catalog No. 502913558
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged L-selectin expressed in CHO cells • Amino acids: 29-332 • MW: 35.6 kDa
Catalog No. 50-291-3558 Supplier Cayman Chemical Supplier No. 35509200 UG
Source: Recombinant human C-terminal His-tagged L-selectin expressed in CHO cells • Amino acids: 29-332 • MW: 35.6 kDa