Learn More
Medchemexpress LLC HY-P0299A 5mg Medchemexpress, LSKL, Inhibitor of Thrombospondin (TSP-1) (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0299A5MG
Encompass_Preferred
Medchemexpress, HY-P0299A 5mg LSKL, Inhibitor of Thrombospondin (TSP-1) (TFA) CAS:0 LSKL, Inhibitor of Thrombospondin (TSP-1) TFA is a latency-associated protein (LAP)-TGFβ derived tetrapeptide and a competitive TGF-β1 antagonist. LSKL, Inhibitor of Thrombospondin (TSP-1) TFA inhibits the binding of TSP-1 to LAP and alleviates renal interstitial fibrosis and hepatic fibrosis. LSKL, Inhibitor of Thrombospondin (TSP-1) TFA suppresses subarachnoid fibrosis via inhibition of TSP-1-mediated TGF-β1 activity, prevents the development of chronic hydrocephalus and improves long-term neurocognitive defects following subarachnoid hemorrhage (SAH). LSKL, Inhibitor of Thrombospondin (TSP-1) TFA can readily crosse the blood-brain barrier[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1658