Learn More
CPC Scientific H-Leu-Ser-Lys-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific THBO001A
Encompass
This peptide, derived from the latency-associated peptide, inhibits thrombospondin (TSP-1) activation of TGF-beta; thus preventing the progression of hepatic damage and fibrosis.SEQUENCE: H-Leu-Ser-Lys-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: LSKL-NH2MOLECULAR FORMULA: C21H42N6O5MOLECULAR WEIGHT: 458.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [283609-79-0]SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Ribeiro, SM. et al. J. Biol. Chem. 274, 13586 (1999)Kondou, H. et al. J. Hepatol. 39, 742 (2003)SKU(s): THBO-001A
Catalog No. 50-211-0558