Learn More
Aviva Systems Biology Corp LSM12 Antibody - middle region (ARP52909_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52909_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSM12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LSM12. Alternate names: FLJ30656, MGC104211, PNAS-135ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60866