Learn More
ABclonal Technology LSM12 Rabbit pAb
SDP
Catalog No. 5000104473
Encompass_Preferred
Nicotinic acid adenine dinucleotide phosphate (NAADP) binding protein (PubMed34362892). Confers NAADP sensitivity to the two pore channel complex (TPCs) by acting as TPC accessory protein necessary for NAADP-evoked Ca(2+) release (PubMed34362892). ( LSM12_HUMAN,Q3MHD2 )
Catalog No. 50-001-04473 Supplier ABclonal Technology Supplier No. A15947500UL
Nicotinic acid adenine dinucleotide phosphate (NAADP) binding protein (PubMed34362892). Confers NAADP sensitivity to the two pore channel complex (TPCs) by acting as TPC accessory protein necessary for NAADP-evoked Ca(2+) release (PubMed34362892). ( LSM12_HUMAN,Q3MHD2 )