Learn More
ABclonal Technology LSM12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A159471000UL
Encompass_Preferred
Nicotinic acid adenine dinucleotide phosphate (NAADP) binding protein (PubMed34362892). Confers NAADP sensitivity to the two pore channel complex (TPCs) by acting as TPC accessory protein necessary for NAADP-evoked Ca(2+) release (PubMed34362892). ( LSM12_HUMAN,Q3MHD2 )
Catalog No. 50-001-07481