Learn More
Aviva Systems Biology Corp LSM2 Antibody - middle region (ARP40963_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40963_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LSM2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human LSM2. Alternate names: G7B, snRNP, C6orf28, YBL026WApplicationIHC, WB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57677