Learn More
Aviva Systems Biology Corp LSM4 Antibody - middle region (ARP40728_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LSM4. Alternate names: YER112W, GRPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53229