Learn More
Aviva Systems Biology Corp LSM6 Antibody - middle region (ARP40708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSM6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LSM6. Alternate names: YDR378CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62880