Learn More
Aviva Systems Biology Corp LSM8 Antibody - N-terminal region (ARP40839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSM8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LSM8. Alternate names: YJR022W, LSM8ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-897