Learn More
Medchemexpress LLC LSP-249 | 1801253-04-2 | 99.1% | 431.92 | 10 MG
SDP
Catalog No. 5000435615
Encompass_Preferred
LSP-249 is a plasma kallikrein inhibitor under study for angioedema. It has an EC50 of less than 100 nM in cell.
  • Plasma kallikrein inhibitor
  • Under study for angioedema
  • EC50 less than 100 nM in cell
  • For research use only
Catalog No. 50-004-35615 Supplier Medchemexpress LLC Supplier No. HY12637810MG
LSP-249 is a plasma kallikrein inhibitor under study for angioedema. It has an EC50 of less than 100 nM in cell.
  • Plasma kallikrein inhibitor
  • Under study for angioedema
  • EC50 less than 100 nM in cell
  • For research use only