Learn More
Medchemexpress LLC LSP-249 | 1801253-04-2 | 99.1% | 431.92 | 100 MG
SDP
Catalog No. 5000437407
Encompass_Preferred
LSP-249 is a plasma kallikrein inhibitor under study for angioedema. It is extracted from patent WO2016011209A1 and has an EC50 less than 100 nM in cell. This product is for research use only and not sold to patients.
  • Plasma kallikrein inhibitor
  • EC50: < 100 nM (kallikrein in cell)
Catalog No. 50-004-37407 Supplier Medchemexpress LLC Supplier No. HY126378100MG
LSP-249 is a plasma kallikrein inhibitor under study for angioedema. It is extracted from patent WO2016011209A1 and has an EC50 less than 100 nM in cell. This product is for research use only and not sold to patients.
  • Plasma kallikrein inhibitor
  • EC50: < 100 nM (kallikrein in cell)