Learn More
Medchemexpress LLC LSP-249 | 1801253-04-2 | 99.1% | 431.92 | 50 MG
SDP
Catalog No. 5000437689
Encompass_Preferred
LSP-249 is a plasma kallikrein inhibitor currently under investigation for its potential use in treating angioedema. This compound was extracted from patent WO2016011209A1 and has demonstrated an EC50 less than 100 nM in cell studies. It is intended for research purposes only and not sold to patients.
  • Plasma kallikrein inhibitor
  • Under study for angioedema
  • EC50 less than 100 nM in cell
Catalog No. 50-004-37689 Supplier Medchemexpress LLC Supplier No. HY12637850MG
LSP-249 is a plasma kallikrein inhibitor currently under investigation for its potential use in treating angioedema. This compound was extracted from patent WO2016011209A1 and has demonstrated an EC50 less than 100 nM in cell studies. It is intended for research purposes only and not sold to patients.
  • Plasma kallikrein inhibitor
  • Under study for angioedema
  • EC50 less than 100 nM in cell