Learn More
Medchemexpress LLC Lsp-249 | 1801253-04-2 | MFCD32671321 | 99.1% | 431.92 g/mol | C24H22ClN5O | 5 MG
SDP
Catalog No. 5000264213
Encompass_Preferred
LSP-249 (CAS 1801253-04-2) is a small-molecule plasma kallikrein inhibitor described in patent literature and provided for preclinical research. It has been reported to show sub-micromolar activity in cell-based assays and is supplied as a well-characterized, high-purity solid for biochemical and cellular testing. Typical packaging is in milligram quantities and material should be handled and stored according to the safety data sheet.
  • High purity suitable for analytical and biological studies.
  • Demonstrated plasma kallikrein inhibitory activity in cell assays.
  • Supplied as a solid, white to light yellow, for easy handling.
  • Available in small milligram packs for preclinical research.
  • Stable under recommended storage for long-term use when stored per SDS.
  • Well-characterized chemical identity: C24H22ClN5O, 431.92 g/mol.
Catalog No. 50-002-64213 Supplier Medchemexpress LLC Supplier No. HY1263785MG
LSP-249 (CAS 1801253-04-2) is a small-molecule plasma kallikrein inhibitor described in patent literature and provided for preclinical research. It has been reported to show sub-micromolar activity in cell-based assays and is supplied as a well-characterized, high-purity solid for biochemical and cellular testing. Typical packaging is in milligram quantities and material should be handled and stored according to the safety data sheet.
  • High purity suitable for analytical and biological studies.
  • Demonstrated plasma kallikrein inhibitory activity in cell assays.
  • Supplied as a solid, white to light yellow, for easy handling.
  • Available in small milligram packs for preclinical research.
  • Stable under recommended storage for long-term use when stored per SDS.
  • Well-characterized chemical identity: C24H22ClN5O, 431.92 g/mol.