Learn More
Sino Biological LSP1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14178T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LSP1 (rh LSP1; Catalog#14178-H08H; AAH01785.1; Met1-Pro339). LSP1 specific IgG was purified by Human LSP1 affinity chromatography.
Catalog No. 50-224-8725