Learn More
Andwin Scientific LSR PNTBL LBEL SHT 3/4I X 2IN
SDP
Catalog No. NC3846720
Encompass_Preferred
LASER PRINTABLE LABEL SHEETS OF LUSTERGUARD METALIZED POLYESTER LABELS 3/4IN X 2IN. 50 PER SHEET FOR 500 SHEETS PER PACK.
Catalog No. NC3846720 Supplier Andwin Scientific Supplier No. LZRATSILVA
LASER PRINTABLE LABEL SHEETS OF LUSTERGUARD METALIZED POLYESTER LABELS 3/4IN X 2IN. 50 PER SHEET FOR 500 SHEETS PER PACK.