Learn More
Aviva Systems Biology Corp LST-3TM12 Antibody - middle region (ARP44578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LST-3TM12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LST-3TM12. Alternate names: LST3, LST-3, LST-3TM12ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-58849