Learn More
Aviva Systems Biology Corp LST1 Antibody - C-terminal region (ARP64242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LST1. It was validated on Western Blot. . Alternate names: B144, D6S49E, LST-1, MGC119006, MGC119007ApplicationWB. Species reactivity: Human
Catalog No. 50-288-212