Learn More
Medchemexpress LLC HY-111486 10mg Medchemexpress, LSZ-102 CAS:2135600-76-7 Purity:>98%
SDP
Catalog No. 501968882
Encompass_Preferred
Medchemexpress, HY-111486 10mg LSZ-102 CAS:2135600-76-7 LSZ-102 is a potent, orally bioavailable selective estrogen receptor degrader with an IC50 of 0.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8882 Supplier Medchemexpress LLC Supplier No. HY11148610MG
Medchemexpress, HY-111486 10mg LSZ-102 CAS:2135600-76-7 LSZ-102 is a potent, orally bioavailable selective estrogen receptor degrader with an IC50 of 0.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.