Learn More
Kuhner Shaker Inc LT-XC WITH HUMIDITY AND CO2 E
Catalog No. 502570035
Encompass
Lab-Therm LT-X with humidityTemperature range: ambient - 15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CBi-directional humidity control to 90%RH, accuracy +/-1%User Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H841 x W855 x D810mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-257-0035 Supplier Kuhner Shaker Inc Supplier No. SMX1703
Add to Cart
Add to Cart
Request a Quote
Lab-Therm LT-X with humidityTemperature range: ambient - 15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CBi-directional humidity control to 90%RH, accuracy +/-1%User Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H841 x W855 x D810mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS