Learn More
Yamato Scientific America, Inc. Yamato Laboratory Stirrer, Max speed of 600 rpm for medium viscosity samples. Complete with stand and rod, stirring shaft and 4-blade propeller. 100V-125V
Catalog No. 501898193 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Laboratory stirrer with well-blanced speed and torque for medium viscosity samples. Highly sensitive feedback system keeps the set speed, even with changing viscosity during stir. Maintenance free DC brushless motor. Digital speed indicator for accurate speed setting and confirmation. Noise prevention measures for optimal work environment. More safety-oriented design. Set includes stand, rod, stainless steel shaft and 60mm 4-blade propeller. Speed range 15~600 rpm.
Catalog No. 50-189-8193 Supplier Yamato Scientific America, Inc. Supplier No. LT400BA
Laboratory stirrer with well-blanced speed and torque for medium viscosity samples. Highly sensitive feedback system keeps the set speed, even with changing viscosity during stir. Maintenance free DC brushless motor. Digital speed indicator for accurate speed setting and confirmation. Noise prevention measures for optimal work environment. More safety-oriented design. Set includes stand, rod, stainless steel shaft and 60mm 4-blade propeller. Speed range 15~600 rpm.