Learn More
Yamato Scientific America, Inc. Yamato Laboratory Stirrer, Max speed of 600 rpm for high viscosity samples. Complete with stand and rod, stirring shaft and 4-blade propeller. 100V-125V
Catalog No. 501898196 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Laboratory stirrer with higher torque for high viscosity samples. Highly sensitive feedback system keeps the set speed, even with changing viscosity during stir. Maintenance free DC brushless motor. Digital speed indicator for accurate speed setting and confirmation. Noise prevention measures for optimal work environment. More safety-oriented design. Set includes stand, rod, stainless steel shaft and 60mm 4-blade propeller. Speed range 15~600 rpm.
Catalog No. 50-189-8196 Supplier Yamato Scientific America, Inc. Supplier No. LT500AA
Laboratory stirrer with higher torque for high viscosity samples. Highly sensitive feedback system keeps the set speed, even with changing viscosity during stir. Maintenance free DC brushless motor. Digital speed indicator for accurate speed setting and confirmation. Noise prevention measures for optimal work environment. More safety-oriented design. Set includes stand, rod, stainless steel shaft and 60mm 4-blade propeller. Speed range 15~600 rpm.