Learn More
Essendant LTA CLOROX GERM. WIPES 420/CS
Catalog No. NC2401447
Encompass_Preferred
Your proven bleach solution for cleaning. Clorox Healthcare Bleach Germicidal Wipes are EPA registered to kill C. difficile and C. auris in three minutes and 58 other microorganisms in one minute or less when used as directed. These one step cleaner disinfectant wipes clean, disinfect and deodorize surfaces with an optimized formula that leaves less residue behind. Clorox Healthcare germicidal wipes have proven efficacy against outbreak causing viruses, emerging viral pathogens, MRSA and VRE. These disinfectant wipes have fast contact times so you can quickly disinfect healthcare surfaces. Use in healthcare settings including hospitals, medical facilities, dental offices, veterinary offices and long-term care facilities. Towel/Wipe Type: Disinfecting Wipes; Application: Hospitals; Medical Facilities; Nursing Homes; Applicable Material: Acrylic; Ceramic; Chrome; Fiberglass; Glass; Metal; Plastic; Tile; Vinyl; Wood; Material(s): Nonwoven Fiber.
Catalog No. NC2401447 Supplier Essendant Supplier No. CLO35309CT
Your proven bleach solution for cleaning. Clorox Healthcare Bleach Germicidal Wipes are EPA registered to kill C. difficile and C. auris in three minutes and 58 other microorganisms in one minute or less when used as directed. These one step cleaner disinfectant wipes clean, disinfect and deodorize surfaces with an optimized formula that leaves less residue behind. Clorox Healthcare germicidal wipes have proven efficacy against outbreak causing viruses, emerging viral pathogens, MRSA and VRE. These disinfectant wipes have fast contact times so you can quickly disinfect healthcare surfaces. Use in healthcare settings including hospitals, medical facilities, dental offices, veterinary offices and long-term care facilities. Towel/Wipe Type: Disinfecting Wipes; Application: Hospitals; Medical Facilities; Nursing Homes; Applicable Material: Acrylic; Ceramic; Chrome; Fiberglass; Glass; Metal; Plastic; Tile; Vinyl; Wood; Material(s): Nonwoven Fiber.