Learn More
Medchemexpress LLC HY-135813 5mg Medchemexpress, LtaS-IN-1 CAS:877950-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1358135MG
Encompass_Preferred
Medchemexpress, HY-135813 5mg LtaS-IN-1 CAS:877950-01-1 LtaS-IN-1 (compound 1771) is a potent small-molecule inhibitor of Lipoteichoic acid (LTA) synthesis in multidrug-resistant (MDR) E. faecium and by altering the cell wall morphology. LtaS-IN-1 alone inhibits Enterococcus.spp 28 strains with varying MIC values ranging from 0.5 μg/mL to 64 μg/mL. LtaS-IN-1 combination with antibiotics abolishs multidrug-resistant E. faecium growth almost completely. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0362