Learn More
Enzo Life Sciences LTB4 ELISA kit (5x96wells)
SDP
Catalog No. 50658221
Encompass_Preferred
The LTB4 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Assay up to 39 samples in duplicate with a broad dynamic range that is suitable for a wide variety of sample matrices. Document your results to scientists world-wide with this highly cited kit. Alternative Name: Leukotriene B4. Sensitivity: 5.63 pg/ml (range 11.7 - 3,000 pg/ml). Assay Time: 4 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-221 Supplier Enzo Life Sciences Supplier No. ADI901068
The LTB4 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Assay up to 39 samples in duplicate with a broad dynamic range that is suitable for a wide variety of sample matrices. Document your results to scientists world-wide with this highly cited kit. Alternative Name: Leukotriene B4. Sensitivity: 5.63 pg/ml (range 11.7 - 3,000 pg/ml). Assay Time: 4 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution