Learn More
Aviva Systems Biology Corp LTB4R2 Antibody - C-terminal region (ARP64274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LTB4R2. It was validated on Western Blot. . Alternate names: BLT2, BLTR2, JULF2, KPG_004, NOP9ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-218