Learn More
AdipoGen anti-LTβR (mouse), mAb (4H8 WH2) (preservative free) (1 mg)
SDP
Supplier:  AdipoGen AG20B0008PFM001
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse LTbetaR. Isotype: Rat IgG2a. Clone: 4H8 WH2. Applications: FACS, FUNC (Blocking). Liquid. In PBS containing 10% glycerol. The LTbetaR activates two different NF-kappaB pathways that lead to distinct patterns of gene induction, including selected chemokines and the cytokine BAFF, which is essential for the survival of mature B lymphocytes. LTbetaR activates the classical NF-kappaB (relA/p50) pathway, like the type 1 TNF receptor (TNFR1), that regulates proinflammatory genes, like the chemokine MIP1beta. However, LTbetaR, unlike TNFR1, also activates the processing of p100 to form RelB/p52 complexes, which activate genes involved in lymphoid organ formation and lymphocyte survival.
Catalog No. 50-205-2902