Learn More
Aviva Systems Biology Corp LTBP1 Antibody - C-terminal region (ARP60287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LTBP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human LTBP1. Alternate names: MGC163161ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-996