Learn More
ABclonal Technology LTBP3 Rabbit pAb
SDP
Catalog No. 5000105784
Encompass_Preferred
The protein encoded by this gene forms a complex with transforming growth factor beta (TGF-beta) proteins and may be involved in their subcellular localization. Activation of this complex requires removal of the encoded binding protein. This protein also may play a structural role in the extracellular matrix. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-05784 Supplier ABclonal Technology Supplier No. A156871000UL
Add to Cart
Add to Cart
The protein encoded by this gene forms a complex with transforming growth factor beta (TGF-beta) proteins and may be involved in their subcellular localization. Activation of this complex requires removal of the encoded binding protein. This protein also may play a structural role in the extracellular matrix. Three transcript variants encoding different isoforms have been found for this gene.