Learn More
Enzo Life Sciences LTbetaR (mouse) monoclonal antibody (5G11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049030100
Encompass_Preferred
Clone: 5G11. Isotype: IgG2a. Immunogen: Extracellular domain of mouse LTβR (lymphotoxin β receptor) expressed in E. coli. Species reactivity: Mouse. Applications: Flow Cytometry. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-0017