Learn More
Enzo Life Sciences LTC4 monoclonal antibody (6E7) (200µg)
SDP
Supplier:  Enzo Life Sciences ADI9059020200
Encompass_Preferred
Clone: 6E7. Host: Mouse. Isotype: IgG. Immunogen: Leukotriene C4. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat. Applications: ELISA Radioimmunoassay. Recommended Dilutions: Radioimmunoassay (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7077