Learn More
Aviva Systems Biology Corp LTC4S Antibody - N-terminal region (ARP49035_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49035_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LTC4S. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LTC4S. Alternate names: MGC33147ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53570