Learn More
MSC Mechanix Wear General Purpose Gloves, Leather, Large, Smooth Grip, 1/Pair
Catalog No. 5000687433
Encompass
These general purpose gloves are constructed from durable grain leather to provide reliable hand protection for tasks ranging from landscaping to home improvement. The smooth grip surface and full-fingered design ensure dexterity and comfort during extended use.
  • Features abrasion and water resistance
  • Equipped with a secure hook and loop cuff style
  • Includes keystone thumb design
  • Full-fingered coverage
  • Durable leather construction with leather lining
  • Meets EN 388 1132X standards
Catalog No. 50-006-87433 Supplier MSC Supplier No. 10409787
These general purpose gloves are constructed from durable grain leather to provide reliable hand protection for tasks ranging from landscaping to home improvement. The smooth grip surface and full-fingered design ensure dexterity and comfort during extended use.
  • Features abrasion and water resistance
  • Equipped with a secure hook and loop cuff style
  • Includes keystone thumb design
  • Full-fingered coverage
  • Durable leather construction with leather lining
  • Meets EN 388 1132X standards