Learn More
Cayman Chemical L-threo-Sphingosine (d18:1), Purity: ≥98%, CAS: 25695-95-8, 5 mg
SDP
Catalog No. 502889592
Encompass_Preferred
An analog of D-erythro-sphingosine; inhibits PKC in mixed micelle assays with 50% inhibition at 2.2 mol % making it a more potent inhibitor compared to D-erythro-sphingosine or other analogs of shorter alkyl chain length
Catalog No. 50-288-9592 Supplier Cayman Chemical Supplier No. 100105415 MG
An analog of D-erythro-sphingosine; inhibits PKC in mixed micelle assays with 50% inhibition at 2.2 mol % making it a more potent inhibitor compared to D-erythro-sphingosine or other analogs of shorter alkyl chain length