Learn More
Selleck Chemical LLC LTI-291 S1024-100mg
SDP
Catalog No. 503108458
Encompass_Preferred
LTI-291 is the first small-molecule activator of Glucocerebrosidase(GCase) for the treatment of parkinson s disease
Catalog No. 50-310-8458 Supplier Selleck Chemical LLC Supplier No. S1024100MG
LTI-291 is the first small-molecule activator of Glucocerebrosidase(GCase) for the treatment of parkinson s disease